Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenoxyacetic acid, 98%, Acros Organics

Click to view available options
Quantity:
25g
1878-87-1
C8H7NO5
197.146
MFCD00016993
TYHHDWAHJRRYCU-UHFFFAOYSA-N
2-nitrophenoxyacetic acid,2-nitrophenoxy acetic acid,2-2-nitrophenoxy acetic acid,o-nitrophenoxyacetic acid,acetic acid, 2-nitrophenoxy,o-nitrophenoxy acetic acid,acmc-209eqj,2-nitrophenoxyaceticacid
15883
CHEBI:66923
C1=CC=C(C(=C1)[N+](=O)[O-])OCC(=O)O
1878-87-1
C8H7NO5
197.146
MFCD00016993
TYHHDWAHJRRYCU-UHFFFAOYSA-N
2-nitrophenoxyacetic acid,2-nitrophenoxy acetic acid,2-2-nitrophenoxy acetic acid,o-nitrophenoxyacetic acid,acetic acid, 2-nitrophenoxy,o-nitrophenoxy acetic acid,acmc-209eqj,2-nitrophenoxyaceticacid
15883
CHEBI:66923
C1=CC=C(C(=C1)[N+](=O)[O-])OCC(=O)O
Infrared Spectrum Authentic
Melting Point 156°C to 160°C
Quantity 25g
Color Beige
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 197.15
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Nitrophenoxyacetic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Risk of explosion if heated under confinement.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning