Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenyl-β-D-galactopyranoside, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
369-07-3
C12H15NO8
301.25
MFCD00063255
KUWPCJHYPSUOFW-YBXAARCKSA-N
2-nitrophenyl-beta-d-galactopyranoside,onpg,2-nitrophenyl beta-d-galactopyranoside,2-nitrophenyl beta-d-galactoside,o-nitrophenyl beta-d-galactopyranoside,o-nitrophenyl-beta-d-galactoside,1-o-o-nitrophenyl-beta-d-galactopyranose,2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy oxane-3,4,5-triol,2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol,2-nitrophenyl b-d-galactopyranoside
96647
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(2-nitrophenoxy)oxane-3,4,5-triol
C1=CC=C(C(=C1)[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
369-07-3
C12H15NO8
301.25
MFCD00063255
KUWPCJHYPSUOFW-YBXAARCKSA-N
2-nitrophenyl-beta-d-galactopyranoside,onpg,2-nitrophenyl beta-d-galactopyranoside,2-nitrophenyl beta-d-galactoside,o-nitrophenyl beta-d-galactopyranoside,o-nitrophenyl-beta-d-galactoside,1-o-o-nitrophenyl-beta-d-galactopyranose,2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy oxane-3,4,5-triol,2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol,2-nitrophenyl b-d-galactopyranoside
96647
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(2-nitrophenoxy)oxane-3,4,5-triol
C1=CC=C(C(=C1)[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
Melting Point 195.0°C
Packaging Glass bottle
Quantity 25g
Specific Rotation − 70.00
Specific Rotation Condition −70° (20°C c=1,H2O)
Color White to Yellow
Infrared Spectrum Authentic
Water 0.2% max. (K.F.)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 301.25
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 2-Nitrophenyl-β-D-galactopyranoside
Show More Show Less