Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenylacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
610-66-2
C8H6N2O2
162.15
MFCD00007183
YPRFCQAWSNWRLM-UHFFFAOYSA-N
2-nitrophenylacetonitrile,2-2-nitrophenyl acetonitrile,2-nitrophenyl acetonitrile,benzeneacetonitrile, 2-nitro,2-nitrobenzyl cyanide,o-nitrophenyl acetonitrile,o-nitrobenzacetonitrile,2-nitrobenzeneacetonitrile,2-nitrobenzyl nitrile,acetonitrile, o-nitrophenyl
11888
2-(2-nitrophenyl)acetonitrile
C1=CC=C(C(=C1)CC#N)[N+](=O)[O-]
610-66-2
C8H6N2O2
162.15
MFCD00007183
YPRFCQAWSNWRLM-UHFFFAOYSA-N
2-nitrophenylacetonitrile,2-2-nitrophenyl acetonitrile,2-nitrophenyl acetonitrile,benzeneacetonitrile, 2-nitro,2-nitrobenzyl cyanide,o-nitrophenyl acetonitrile,o-nitrobenzacetonitrile,2-nitrobenzeneacetonitrile,2-nitrobenzyl nitrile,acetonitrile, o-nitrophenyl
11888
2-(2-nitrophenyl)acetonitrile
C1=CC=C(C(=C1)CC#N)[N+](=O)[O-]
Quantity 50g
Melting Point 81.0°C to 85.0°C
Color Brown to Brown-Gray
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula O2NC6H4CH2CN
Beilstein 09, 455
Formula Weight 162.15
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Nitrophenylacetonitrile
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning

EINECSNumber : 210-231-