Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-nitrophenylarsonic acid, +90%, ACROS Organics™

Click to view available options
Quantity:
1g
5410-29-7
C6H6AsNO5
247.038
MFCD00047664
UYEDGVZDGVIURN-UHFFFAOYSA-N
2-nitrobenzenearsonic acid,arsonic acid, 2-nitrophenyl,o-nitrophenylarsenic acid,o-nitrobenzenearsonic acid,o-nitrophenyl arsonic acid,benzenearsonic acid, o-nitro,unii-hnr3su2y5p,2-nitrophenyl arsonic acid,arsonic acid, as-2-nitrophenyl,hnr3su2y5p
79424
(2-nitrophenyl)arsonic acid
C1=CC=C(C(=C1)[N+](=O)[O-])[As](=O)(O)O
5410-29-7
C6H6AsNO5
247.038
MFCD00047664
UYEDGVZDGVIURN-UHFFFAOYSA-N
2-nitrobenzenearsonic acid,arsonic acid, 2-nitrophenyl,o-nitrophenylarsenic acid,o-nitrobenzenearsonic acid,o-nitrophenyl arsonic acid,benzenearsonic acid, o-nitro,unii-hnr3su2y5p,2-nitrophenyl arsonic acid,arsonic acid, as-2-nitrophenyl,hnr3su2y5p
79424
(2-nitrophenyl)arsonic acid
C1=CC=C(C(=C1)[N+](=O)[O-])[As](=O)(O)O
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 1g
Color Beige-Yellow
Melting Point 225.0°C
CAS Min % 90.0
CAS Max % 100.0
Assay Percent Range 90+%
Linear Formula NO2C6H4AsO3H2
Solubility Information Solubility in water: 2.5g/100mL.
Formula Weight 247.04
Physical Form Powder
Percent Purity 90+%
Chemical Name or Material 2-Nitrophenylarsonic acid
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger