Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
190788-59-1
C12H16BNO4
249.08
VLJYUDGCEKORNG-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-2-nitrophenyl-1,3,2-dioxaborolane,2-nitrobenzeneboronic acid pinacol ester,2-nitrophenyl boronic acid pinacol ester,2-nitrophenylboronic acid pinacol ester,1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-nitrophenyl,2-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene,amtb916,2-nitrophenyl boronic acid, pinacol ester,2-nitrophenylboronic acid, pinacol ester
3699568
4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2[N+](=O)[O-]
190788-59-1
C12H16BNO4
249.08
VLJYUDGCEKORNG-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-2-nitrophenyl-1,3,2-dioxaborolane,2-nitrobenzeneboronic acid pinacol ester,2-nitrophenyl boronic acid pinacol ester,2-nitrophenylboronic acid pinacol ester,1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-nitrophenyl,2-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene,amtb916,2-nitrophenyl boronic acid, pinacol ester,2-nitrophenylboronic acid, pinacol ester
3699568
4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2[N+](=O)[O-]
Quantity 5g
Melting Point 45.0°C to 47.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 249.08
Percent Purity 97%
Chemical Name or Material 2-Nitrophenylboronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning