Alfa Aesar™ 2-Nitroso-1-naphthol-4-sulfonic acid tetrahydrate, 94%

Manufacturer: Alfa Aesar™ B2459314

Quantity 25g

 View more versions of this product

Catalog No. AAB2459314

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 624725-88-8
    Molecular Formula C10H7NO5S·4H2O
    Formula Weight 325.30 (253.24 Anhydrous)
    MDL Number MFCD02682113
    Beilstein 2136360
    Chemical Name or Material 2-Nitroso-1-naphthol-4-sulfonic acid tetrahydrate

    Specifications

    Formula Weight 325.30 (253.24 Anhydrous)
    Molecular Formula C10H7NO5S·4H2O
    MDL Number MFCD02682113
    Quantity 25g
    TSCA TSCA
    CAS 624725-88-8
    Beilstein 2136360
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈143°C (dec.)
    Chemical Name or Material 2-Nitroso-1-naphthol-4-sulfonic acid tetrahydrate
    Assay 94%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit