Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nonanone, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081696
Click to view available options
Quantity:
50 g
100 g
1000 g
821-55-6
C9H18O
142.242
MFCD00009553
VKCYHJWLYTUGCC-UHFFFAOYSA-N
2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
13187
CHEBI:77927
nonan-2-one
CCCCCCCC(=O)C
821-55-6
C9H18O
142.242
MFCD00009553
VKCYHJWLYTUGCC-UHFFFAOYSA-N
2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
13187
CHEBI:77927
nonan-2-one
CCCCCCCC(=O)C

2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point -15°C
Density 0.826
Boiling Point 195°C
Flash Point 64°C (147°F)
Refractive Index 1.421
Quantity 50 g
Beilstein 1743645
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 142.24
Percent Purity ≥98%
Chemical Name or Material 2-Nonanone
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-480-0
RTECSNumber RA8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit