Alfa Aesar™ 2-Nonyne, 98%

Manufacturer: Alfa Aesar™ B2362006

Quantity 5g

 View more versions of this product

Catalog No. AAB2362006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19447-29-1
    Molecular Formula C9H16
    Linear Formula CH3C≡C(CH2)5CH3
    Formula Weight 124.23
    MDL Number MFCD00039986
    UN Number UN3295
    Synonym Methyl n-hexyl acetylene
    Chemical Name or Material 2-Nonyne

    Specifications

    Formula Weight 124.23
    Quantity 5g
    Molecular Formula C9H16
    Linear Formula CH3C≡C(CH2)5CH3
    MDL Number MFCD00039986
    CAS 19447-29-1
    UN Number UN3295
    Synonym Methyl n-hexyl acetylene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P301+P330+P331-P315
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Boiling Point 158° to 159°C
    Chemical Name or Material 2-Nonyne
    Assay 98%

    GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapour.

    GHS P Statement
    P301+P330+P331-P315
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit