Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Nonyne, 98%

Click to view available options
Quantity:
5g
25g
19447-29-1
C9H16
124.227
MFCD00039986
LXKRETAGISZJAD-UHFFFAOYSA-N
2-nonyne,acmc-1bt6l
140536
non-2-yne
CCCCCCC#CC
19447-29-1
C9H16
124.227
MFCD00039986
LXKRETAGISZJAD-UHFFFAOYSA-N
2-nonyne,acmc-1bt6l
140536
non-2-yne
CCCCCCC#CC
Quantity 25g
Boiling Point 158°C to 159°C
Linear Formula CH3C≡C(CH2)5CH3
UN Number UN3295
Formula Weight 124.23
Percent Purity 98%
Chemical Name or Material 2-Nonyne

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.

GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.