Alfa Aesar™ 2-Norbornene-2-boronic acid

Manufacturer: Alfa Aesar™ H53221MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH53221MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 871333-98-1
    Molecular Formula C7H11BO2
    Formula Weight 137.97
    MDL Number MFCD07783853
    Synonym Bicyclo[2.2.1]hept-2-en-2-ylboronic acid, 95%
    Chemical Name or Material 2-Norbornene-2-boronic acid

    Specifications

    Formula Weight 137.97
    Molecular Formula C7H11BO2
    MDL Number MFCD07783853
    CAS 871333-98-1
    Synonym Bicyclo[2.2.1]hept-2-en-2-ylboronic acid, 95%
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 124° to 126°C
    Quantity 250mg
    Chemical Name or Material 2-Norbornene-2-boronic acid

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit