Alfa Aesar™ 2-Octanone, 98%

Manufacturer: Alfa Aesar™ A11643AU

Quantity 1000mL

 View more versions of this product

Catalog No. AAA11643AU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 111-13-7
    Molecular Formula CH3(CH2)5C(O)CH3
    Formula Weight 128.22
    MDL Number MFCD00009540
    Beilstein 635843
    Merck Index 14,4711
    Synonym n-Hexyl methyl ketone
    Chemical Name or Material 2-Octanone

    Specifications

    Formula Weight 128.22
    Health Hazard 3 GHS P Statement
    P210-P261-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 111-13-7
    Health Hazard 1 Danger
    Boiling Point 172° to 173°C
    Flash Point 63°C (145°F)
    RTECS Number RH1484000
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H311-H227-H315-H319-H335
    Toxic in contact with skin.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula CH3(CH2)5C(O)CH3
    Merck Index 14,4711
    Synonym n-Hexyl methyl ketone
    Melting Point −16°C
    Quantity 1000mL
    MDL Number MFCD00009540
    Beilstein 635843
    Chemical Name or Material 2-Octanone
    Assay 98%

    Description

    Applications
    2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

    Solubility
    Slightly soluble in water.

    Notes
    Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

    GHS H Statement
    H311-H227-H315-H319-H335
    Toxic in contact with skin.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit