Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octanone, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
500mL
2.5L
Packaging:
Glass bottle
111-13-7
C8H16O
128.21
MFCD00009540
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
2-octanone,n-hexyl methyl ketone,hexyl methyl ketone,methyl hexyl ketone,methyl n-hexyl ketone,octanone,2-oxooctane,2-octanone natural,fema number 2802,unii-j2g84h29af
8093
CHEBI:87434
octan-2-one
CCCCCCC(=O)C
111-13-7
C8H16O
128.21
MFCD00009540
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
2-octanone,n-hexyl methyl ketone,hexyl methyl ketone,methyl hexyl ketone,methyl n-hexyl ketone,octanone,2-oxooctane,2-octanone natural,fema number 2802,unii-j2g84h29af
8093
CHEBI:87434
octan-2-one
CCCCCCC(=O)C
Melting Point -16°C
Boiling Point 173°C
Flash Point 55°C
Specific Gravity 0.819
Refractive Index 1.4140 to 1.416
Packaging Glass bottle
Color Colorless to Yellow
Quantity 2.5L
Infrared Spectrum Authentic
Density 0.8190g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3(CH2)5COCH3
Beilstein 01, 704
Merck Index 15, 4747
Solubility Information Solubility in water: 0.9g/L. Other solubilities: miscible with alcohol,ether,hydrocarbons and,esters
Formula Weight 128.21
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material 2-Octanone, 99+%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-837-1

RTECSNumber : RH1484000

TSCA : TSCA