Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octanone oxime, 99%, Acros Organics

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
7207-49-0
C8H17NO
143.23
MFCD00089167
GZRPVYSKBVDCBV-CMDGGOBGSA-N
2-octanone, oxime,2-octanone oxime,n-octan-2-ylidene hydroxylamine,2-hydroxyiminooctane,z-n-octan-2-ylidene hydroxylamine
9562584
(NE)-N-octan-2-ylidenehydroxylamine
CCCCCCC(=NO)C
7207-49-0
C8H17NO
143.23
MFCD00089167
GZRPVYSKBVDCBV-CMDGGOBGSA-N
2-octanone, oxime,2-octanone oxime,n-octan-2-ylidene hydroxylamine,2-hydroxyiminooctane,z-n-octan-2-ylidene hydroxylamine
9562584
(NE)-N-octan-2-ylidenehydroxylamine
CCCCCCC(=NO)C
Infrared Spectrum Authentic
Packaging Glass bottle
Density 0.8800g/mL
Refractive Index 1.4510 to 1.453
Specific Gravity 0.88
Quantity 100g
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)5C(CH3)=NOH
Formula Weight 143.23
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Octanone oxime
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-586-5