Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octyl-1-dodecanol, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
5333-42-6
5333-42-6
CAS Max % 1
Packaging Poly Drum
Quantity 20 L
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.