Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25mL
693-16-3
C8H19N
129.247
HBXNJMZWGSCKPW-UHFFFAOYSA-N
2-aminooctane,2-octanamine,2-octylamine,1-methylheptylamine,heptylamine, 1-methyl,2-caprylamine,sec-octylamine,1-methyl heptylamine,oct-2-ylamine,sec-octanamine
12735
octan-2-amine
CCCCCCC(C)N
693-16-3
C8H19N
129.247
HBXNJMZWGSCKPW-UHFFFAOYSA-N
2-aminooctane,2-octanamine,2-octylamine,1-methylheptylamine,heptylamine, 1-methyl,2-caprylamine,sec-octylamine,1-methyl heptylamine,oct-2-ylamine,sec-octanamine
12735
octan-2-amine
CCCCCCC(C)N
Density 0.771g/mL
Refractive Index 1.4225 to 1.4245 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 10g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)5CH(CH3)NH2
Beilstein 04, 196
Formula Weight 129.24
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Octylamine, 98%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-744-2

RTECSNumber : MK1210000

TSCA : TSCA