Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octyn-1-ol, 98%, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
20739-58-6
C8H14O
126.2
MFCD00039542
TTWYFVOMGMBZCF-UHFFFAOYSA-N
2-octyn-1-ol,2-octyne-1-ol,pubchem13066,acmc-1clfg,2-octyn 1-ol,2-octyn-1-ol 10g
140750
oct-2-yn-1-ol
CCCCCC#CCO
20739-58-6
C8H14O
126.2
MFCD00039542
TTWYFVOMGMBZCF-UHFFFAOYSA-N
2-octyn-1-ol,2-octyne-1-ol,pubchem13066,acmc-1clfg,2-octyn 1-ol,2-octyn-1-ol 10g
140750
oct-2-yn-1-ol
CCCCCC#CCO
Boiling Point 76°C to 78°C (2.0mmHg)
Specific Gravity 0.88
Flash Point 91°C
Refractive Index 1.4550 to 1.457
Density 0.8800g/mL
Infrared Spectrum Authentic
Quantity 25mL
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 126.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Octyn-1-ol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning