Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octyne, 97%, stabilized, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
2809-67-8
C8H14
110.2
QCQALVMFTWRCFI-UHFFFAOYSA-N
2-octyne,n-c5h11c.$.cch3,amylmethylacetylene,methylpentylacetylene,acmc-209h0u,n-c5h11c.equiv.cch3
17769
oct-2-yne
CCCCCC#CC
2809-67-8
C8H14
110.2
QCQALVMFTWRCFI-UHFFFAOYSA-N
2-octyne,n-c5h11c.$.cch3,amylmethylacetylene,methylpentylacetylene,acmc-209h0u,n-c5h11c.equiv.cch3
17769
oct-2-yne
CCCCCC#CC
Infrared Spectrum Authentic
Flash Point 18°C
Quantity 100g
Specific Gravity 0.759
Stabilizer Approx. 200ppm
Boiling Point 137.0°C
Density 0.7590g/mL
Assay Percent Range 96% min. (GC)
Formula Weight 110.2
Percent Purity 97%
Chemical Name or Material 2-Octyne
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

GHS Signal Word: Danger

EINECSNumber : 220-553-3