Alfa Aesar™ 2-Octyne, 98+%

Manufacturer: Alfa Aesar™ A1019730

Quantity 250g

 View more versions of this product

Catalog No. AAA1019730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2809-67-8
    Molecular Formula CH3(CH2)4C≡CCH3
    Formula Weight 110.2
    MDL Number MFCD00027302
    UN Number UN3295
    Beilstein 1697689
    Synonym n-Pentyl methyl acetylene
    Chemical Name or Material 2-Octyne

    Specifications

    Formula Weight 110.2
    Molecular Formula CH3(CH2)4C≡CCH3
    CAS 2809-67-8
    Health Hazard 1 Danger
    Quantity 250g
    MDL Number MFCD00027302
    UN Number UN3295
    Beilstein 1697689
    Synonym n-Pentyl methyl acetylene
    Health Hazard 2 GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    Flash Point 28°C (82°F)
    Health Hazard 3 GHS P Statement
    P210-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.
    Boiling Point 137° to 138°C
    Odor Petroleum-like
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Chemical Name or Material 2-Octyne
    Assay ≥98%

    GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit