Learn More
2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%, Thermo Scientific Chemicals
Description
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.
Notes
Hygroscopic. Store at room temperature.
Specifications
Melting Point | 208°C |
Color | White to Gray |
Density | 1.048 g/mL |
Quantity | 250 mg |
Formula Weight | 172.2 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 2-Oxa-7-azaspiro[3.5]nonane hemioxalate |
Safety and Handling
Hazard Category | H302+H312 |
Hazard Statement | GHS H Statement H302-H312 Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c |
TSCA | No |
storageNote1 | 2° to 8°C |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.