Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxazolidone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
497-25-6
C3H5NO2
87.08
MFCD00005268
IZXIZTKNFFYFOF-UHFFFAOYSA-N
2-oxazolidone,oxazolidin-2-one,2-oxazolidinone,oxazolidinone,oxazolidone,oxazolidinones,unii-z4d49w92pp,carbamic acid, 2-hydroxyethyl-, gamma-lactone,wln: t5mvotj,carbamic acid, .gamma.-lactone
73949
CHEBI:1237
1,3-oxazolidin-2-one
C1COC(=O)N1
497-25-6
C3H5NO2
87.08
MFCD00005268
IZXIZTKNFFYFOF-UHFFFAOYSA-N
2-oxazolidone,oxazolidin-2-one,2-oxazolidinone,oxazolidinone,oxazolidone,oxazolidinones,unii-z4d49w92pp,carbamic acid, 2-hydroxyethyl-, gamma-lactone,wln: t5mvotj,carbamic acid, .gamma.-lactone
73949
CHEBI:1237
1,3-oxazolidin-2-one
C1COC(=O)N1
Boiling Point 220°C (48.0mmHg)
Melting Point 86°C to 89°C
Color Beige-Yellow to White
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 27,135
Fieser 12,359; 13,24; 14,242
Solubility Information Solubility in water: soluble
Formula Weight 87.08
Physical Form Crystals or Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Oxazolidone, 98%
Show More Show Less

GHS H Statement:
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 207-840-9

RTECSNumber : RQ2450000

TSCA : TSCA