Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Oxetanemethanol, 96%

Click to view available options
Quantity:
250mg
1g
61266-70-4
C4H8O2
88.106
MFCD00142933
PQZJTHGEFIQMCO-UHFFFAOYSA-N
2-hydroxymethyloxetane,2-oxetanemethanol,2-oxetanylmethanol,2,4-epoxybutanol,oxetan-2-yl-methanol,oxetane-2-methanol,methyloloxacyclobutan,2-hydrpxymethyloxetane,oxetan-2-yl methanol,rac-2-hydroxymethyloxetane
12284142
oxetan-2-ylmethanol
C1COC1CO
61266-70-4
C4H8O2
88.106
MFCD00142933
PQZJTHGEFIQMCO-UHFFFAOYSA-N
2-hydroxymethyloxetane,2-oxetanemethanol,2-oxetanylmethanol,2,4-epoxybutanol,oxetan-2-yl-methanol,oxetane-2-methanol,methyloloxacyclobutan,2-hydrpxymethyloxetane,oxetan-2-yl methanol,rac-2-hydroxymethyloxetane
12284142
oxetan-2-ylmethanol
C1COC1CO
Applications
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in freezer at (-20°C). Store away form water/moisture. Store away from oxidizing agent.
Density 1.080g/mL
Color Colorless to Yellow
Quantity 250mg
Boiling Point 98°C (15mmHg)
Sensitivity Moisture Sensitive
Refractive Index 1.4460
Formula Weight 88.11
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 2-Oxetanemethanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C