Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(p-Tolyl)propionic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
938-94-3
C10H12O2
164.204
MFCD01111378
KDYOFXPLHVSIHS-UHFFFAOYSA-N
2-4-methylphenyl propanoic acid,2-4-methylphenyl propionic acid,2-p-tolyl propanoic acid,p-methylhydratropic acid,propanoic acid, 2-4-methylphenyl,2rs-2-4-methylphenyl propanoic acid,ibuprofen impurity d,alpha,4-dimethylbenzeneacetic acid,2-4-tolyl propanoic acid,acmc-209rnz
150866
2-(4-methylphenyl)propanoic acid
CC1=CC=C(C=C1)C(C)C(=O)O
938-94-3
C10H12O2
164.204
MFCD01111378
KDYOFXPLHVSIHS-UHFFFAOYSA-N
2-4-methylphenyl propanoic acid,2-4-methylphenyl propionic acid,2-p-tolyl propanoic acid,p-methylhydratropic acid,propanoic acid, 2-4-methylphenyl,2rs-2-4-methylphenyl propanoic acid,ibuprofen impurity d,alpha,4-dimethylbenzeneacetic acid,2-4-tolyl propanoic acid,acmc-209rnz
150866
2-(4-methylphenyl)propanoic acid
CC1=CC=C(C=C1)C(C)C(=O)O
Quantity 5g
Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOOCCH(CH3)C6H4CH3
Beilstein 09, II, 360
Formula Weight 164.2
Physical Form Liquid After Melting
Percent Purity ≥97.5%
Chemical Name or Material 2-(p-Tolyl)propionic acid, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA