Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
Packaging:
Glass bottle
2688-84-8
C12H11NO
185.22
MFCD00035765
NMFFUUFPJJOWHK-UHFFFAOYSA-N
2-aminodiphenyl ether,o-phenoxyaniline,2-aminophenyl phenyl ether,benzenamine, 2-phenoxy,2-ade,2-phenoxybenzenamine,o-aminophenyl phenyl ether,aniline, 2-phenoxy,aniline, o-phenoxy,2-phonoxyaniline
75899
2-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=CC=C2N
2688-84-8
C12H11NO
185.22
MFCD00035765
NMFFUUFPJJOWHK-UHFFFAOYSA-N
2-aminodiphenyl ether,o-phenoxyaniline,2-aminophenyl phenyl ether,benzenamine, 2-phenoxy,2-ade,2-phenoxybenzenamine,o-aminophenyl phenyl ether,aniline, 2-phenoxy,aniline, o-phenoxy,2-phonoxyaniline
75899
2-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=CC=C2N
Boiling Point 170°C (18.0mmHg)
Flash Point 113°C
Melting Point 46°C to 50°C
Color Brown to Gray
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5OC6H4NH2
Solubility Information Solubility in water: insoluble
Formula Weight 185.22
Physical Form Crystalline Powder or Flakes
Percent Purity ≥97.5%
Chemical Name or Material 2-Phenoxyaniline
Show More Show Less

GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning