Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
Packaging:
Glass bottle
2688-84-8
C12H11NO
185.22
MFCD00035765
NMFFUUFPJJOWHK-UHFFFAOYSA-N
2-aminodiphenyl ether,o-phenoxyaniline,2-aminophenyl phenyl ether,benzenamine, 2-phenoxy,2-ade,2-phenoxybenzenamine,o-aminophenyl phenyl ether,aniline, 2-phenoxy,aniline, o-phenoxy,2-phonoxyaniline
75899
2-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=CC=C2N
2688-84-8
C12H11NO
185.22
MFCD00035765
NMFFUUFPJJOWHK-UHFFFAOYSA-N
2-aminodiphenyl ether,o-phenoxyaniline,2-aminophenyl phenyl ether,benzenamine, 2-phenoxy,2-ade,2-phenoxybenzenamine,o-aminophenyl phenyl ether,aniline, 2-phenoxy,aniline, o-phenoxy,2-phonoxyaniline
75899
2-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=CC=C2N
Boiling Point 170°C (18.0mmHg)
Flash Point 113°C
Packaging Glass bottle
Color Brown to Gray
Infrared Spectrum Authentic
Melting Point 46°C to 50°C
Quantity 500g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5OC6H4NH2
Solubility Information Solubility in water: insoluble
Formula Weight 185.22
Physical Form Crystalline Powder or Flakes
Percent Purity 98%
Chemical Name or Material 2-Phenoxyaniline
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 220-254-8