Alfa Aesar™ 2-Phenoxyethanol, 94%

Manufacturer: Alfa Aesar™ A1078630

Quantity 250g

 View more versions of this product

Catalog No. AAA1078630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122-99-6
    Molecular Formula C8H10O2
    Linear Formula C6H5O(CH2)2OH
    Formula Weight 138.17
    MDL Number MFCD00002857
    Beilstein 1364011
    Merck Index 14,7257
    Synonym Ethylene glycol monophenyl ether
    Chemical Name or Material 2-Phenoxyethanol

    Specifications

    Formula Weight 138.17
    Melting Point 11° to 14°C
    Assay Percent Range 94%
    Molecular Formula C8H10O2
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.5370
    Physical Form Liquid
    Merck Index 14,7257
    Health Hazard 1 Warning
    Boiling Point 245°C
    Color Colorless
    Quantity 250g
    TSCA TSCA
    MDL Number MFCD00002857
    CAS 122-99-6
    Beilstein 1364011
    Health Hazard 2 GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.
    RTECS Number KM0350000
    Density 1.102g/mL
    Linear Formula C6H5O(CH2)2OH
    Synonym Ethylene glycol monophenyl ether
    Chemical Name or Material 2-Phenoxyethanol
    Assay 94%

    Description

    Applications
    2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.

    Solubility
    Miscible with water, ethanol, propanol and propylene glycol.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit