Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyethanol, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

122-99-6
C8H10O2
138.166
QCDWFXQBSFUVSP-UHFFFAOYSA-N
31236
CHEBI:64275
2-phenoxyethanol
C1=CC=C(C=C1)OCCO

A grade of chemicals for laboratory use

Flash Point 121.00°C
Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.11kg
Formula Weight 138.17
Chemical Name or Material 2-Phenoxyethanol