Alfa Aesar™ 2-Phenoxyethylamine, 98%

Manufacturer: Alfa Aesar™ A1222503

Quantity 1g

 View more versions of this product

Catalog No. AAA1222503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1758-46-9
    Molecular Formula C8H11NO
    Linear Formula C6H5O(CH2)2NH2
    Formula Weight 137.18
    MDL Number MFCD00052975
    UN Number UN2735
    Beilstein 774671
    Synonym 1-Amino-2-phenoxyethane
    Chemical Name or Material 2-Phenoxyethylamine

    Specifications

    Formula Weight 137.18
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P308+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.
    Synonym 1-Amino-2-phenoxyethane
    RTECS Number KR9500000
    Flash Point 107°C (224°F)
    MDL Number MFCD00052975
    CAS 1758-46-9
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Odor Amine-like
    Quantity 1g
    Molecular Formula C8H11NO
    UN Number UN2735
    Health Hazard 1 Danger
    Boiling Point 88° to 90°C (1mmHg)
    Linear Formula C6H5O(CH2)2NH2
    Beilstein 774671
    Chemical Name or Material 2-Phenoxyethylamine
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P308+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit