Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Phenoxymethyl)benzoic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
724-98-1
C14H12O3
228.247
MFCD00020294
YKNORODREYVARM-UHFFFAOYSA-N
2-phenoxymethyl benzoic acid,benzoic acid, 2-phenoxymethyl,alpha-phenoxy-o-toluic acid,unii-nw1f23eg53,o-phenoxymethyl benzoic acid,phenoxymethylbenzoic acid,acmc-1bgx0
69761
2-(phenoxymethyl)benzoic acid
C1=CC=C(C=C1)OCC2=CC=CC=C2C(=O)O
724-98-1
C14H12O3
228.247
MFCD00020294
YKNORODREYVARM-UHFFFAOYSA-N
2-phenoxymethyl benzoic acid,benzoic acid, 2-phenoxymethyl,alpha-phenoxy-o-toluic acid,unii-nw1f23eg53,o-phenoxymethyl benzoic acid,phenoxymethylbenzoic acid,acmc-1bgx0
69761
2-(phenoxymethyl)benzoic acid
C1=CC=C(C=C1)OCC2=CC=CC=C2C(=O)O
Quantity 1g
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 2-(Phenoxymethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA