Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-1,3-propanediol, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
1570-95-2
C9H12O2
152.19
BPBDZXFJDMJLIB-UHFFFAOYSA-N
2-phenyl-1,3-propanediol,2-phenyl-1,3-propanediole,2-phenyl-propane-1,3-diol,2-phenyl-1,3-propane diol,unii-9f93674jbb,1,3-propanediol, 2-phenyl,phenylpropanediol,pubchem20386,rarechem al bd 1370,acmc-1bz5a
254178
2-phenylpropane-1,3-diol
C1=CC=C(C=C1)C(CO)CO
1570-95-2
C9H12O2
152.19
BPBDZXFJDMJLIB-UHFFFAOYSA-N
2-phenyl-1,3-propanediol,2-phenyl-1,3-propanediole,2-phenyl-propane-1,3-diol,2-phenyl-1,3-propane diol,unii-9f93674jbb,1,3-propanediol, 2-phenyl,phenylpropanediol,pubchem20386,rarechem al bd 1370,acmc-1bz5a
254178
2-phenylpropane-1,3-diol
C1=CC=C(C=C1)C(CO)CO
Melting Point 54.0°C to 56.0°C
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-1,3-propanediol

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger