Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-2-imidazoline, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
250g
500g
Packaging:
Glass bottle
936-49-2
C9H10N2
146.19
MFCD00005180
BKCCAYLNRIRKDJ-UHFFFAOYSA-N
2-phenyl-2-imidazoline,2-phenylimidazoline,1h-imidazole, 4,5-dihydro-2-phenyl,2-imidazoline, 2-phenyl,4,5-dihydro-2-phenyl-1h-imidazole,2-phenyl imidazoline,2-phenyl-2-imidazolin,acmc-209rmt,labotest-bb lt00000058,5-23-06-00425 beilstein handbook reference
13639
2-phenyl-4,5-dihydro-1H-imidazole
C1CN=C(N1)C2=CC=CC=C2
936-49-2
C9H10N2
146.19
MFCD00005180
BKCCAYLNRIRKDJ-UHFFFAOYSA-N
2-phenyl-2-imidazoline,2-phenylimidazoline,1h-imidazole, 4,5-dihydro-2-phenyl,2-imidazoline, 2-phenyl,4,5-dihydro-2-phenyl-1h-imidazole,2-phenyl imidazoline,2-phenyl-2-imidazolin,acmc-209rmt,labotest-bb lt00000058,5-23-06-00425 beilstein handbook reference
13639
2-phenyl-4,5-dihydro-1H-imidazole
C1CN=C(N1)C2=CC=CC=C2
Melting Point 94°C to 99°C
Boiling Point 188°C (20.0mmHg)
Flash Point 201°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Beilstein 23, V,6, 425
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 146.19
Physical Form Flakes or Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 2-Phenyl-2-imidazoline, 98+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-313-4

RTECSNumber : NJ4395500

TSCA : TSCA