Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-3-butyn-2-ol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
127-66-2
C10H10O
146.19
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O
127-66-2
C10H10O
146.19
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O
Flash Point 96°C
Quantity 25g
Infrared Spectrum Authentic
Boiling Point 102.0°C to 103.0°C (12.0 mmHg)
Packaging Glass bottle
Melting Point 50.0°C to 52.0°C
Assay Percent Range 98% min. (GC)
Linear Formula HC≡CC(C6H5)(OH)CH3
Beilstein 06,II,559
Merck Index 10,7158
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone, benzene and most other common, organic solvents
Formula Weight 146.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-3-butyn-2-ol
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
Do

GHS Signal Word: Warning

EINECSNumber : 204-855-2