Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-3-butyn-2-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-7077194
Click to view available options
Quantity:
5 g
25 g
127-66-2
C10H10O
146.19
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O
127-66-2
C10H10O
146.19
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 50.0°C to 52.0°C
Boiling Point 102.0°C to 103.0°C (12.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CC(C6H5)(OH)CH3
Quantity 5 g
Beilstein 06,II,559
Merck Index 10,7158
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone, benzene and most other common, organic solvents
Formula Weight 146.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-3-butyn-2-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
Do
EINECSNumber 204-855-2

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit