Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Phenyl-3-butyn-2-ol, 98%

Click to view available options
Quantity:
5g
25g
100g
127-66-2
C10H10O
146.189
MFCD00004454
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol,methylphenylethynylcarbinol,3-hydroxy-3-phenyl-1-butyne,3-butyn-2-ol, 2-phenyl,3-phenyl-butin-1-ol-3,alpha-ethynyl-alpha-methylbenzyl alcohol,alpha-ethynyl-alpha-methylbenzenemethanol,benzenemethanol, .alpha.-ethynyl-.alpha.-methyl,3-phenyl-butin-1-ol-3 german,benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O
127-66-2
C10H10O
146.189
MFCD00004454
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol,methylphenylethynylcarbinol,3-hydroxy-3-phenyl-1-butyne,3-butyn-2-ol, 2-phenyl,3-phenyl-butin-1-ol-3,alpha-ethynyl-alpha-methylbenzyl alcohol,alpha-ethynyl-alpha-methylbenzenemethanol,benzenemethanol, .alpha.-ethynyl-.alpha.-methyl,3-phenyl-butin-1-ol-3 german,benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O
Boiling Point 92°C to 93°C (10mmHg)
Flash Point 96°C (205°F)
Melting Point 49°C to 53°C
Quantity 5g
Beilstein 1100096
Formula Weight 146.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-3-butyn-2-ol

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : ES0840000

TSCA : TSCA