Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-4-quinolinecarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
132-60-5
C16H11NO2
249.269
MFCD00006750
YTRMTPPVNRALON-UHFFFAOYSA-N
cinchophen,tervalon,2-phenyl-4-quinolinecarboxylic acid,cinchophene,cinconal,phenoquin,quinofen,quinophan,quinophen,atofan
8593
2-phenylquinoline-4-carboxylic acid
C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)C(=O)O
132-60-5
C16H11NO2
249.269
MFCD00006750
YTRMTPPVNRALON-UHFFFAOYSA-N
cinchophen,tervalon,2-phenyl-4-quinolinecarboxylic acid,cinchophene,cinconal,phenoquin,quinofen,quinophan,quinophen,atofan
8593
2-phenylquinoline-4-carboxylic acid
C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)C(=O)O
Quantity 25g
Infrared Spectrum Authentic
Melting Point 213°C to 216°C
Color Beige
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 22, 103
Merck Index 15, 2291
Formula Weight 249.27
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Phenyl-4-quinolinecarboxylic acid, 99%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-067-1

RTECSNumber : GD4025000

TSCA : TSCA