Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Phenyl-L-glycinamide hydrochloride, 97%

Click to view available options
Quantity:
1g
5g
60079-51-8
C8H11ClN2O
186.639
MFCD00270568
LXOAFHGMXCFTOU-FJXQXJEOSA-N
s-2-amino-2-phenylacetamide hydrochloride,l-phenylglycine amide hydrochloride,h-phg-nh2 hcl,h-phg-nh2.hcl,2s-2-amino-2-phenylacetamide hydrochloride,hpphg-nh2 hcl,h-phg-nh2 . hcl,phenylglycine amide hydrochloride,s-2-amino-2-phenyl-acetamide hydrochloride,alphas-alpha-amino-benzeneacetamide hydrochloride
22850726
(2S)-2-amino-2-phenylacetamide;hydrochloride
C1=CC=C(C=C1)C(C(=O)N)N.Cl
60079-51-8
C8H11ClN2O
186.639
MFCD00270568
LXOAFHGMXCFTOU-FJXQXJEOSA-N
s-2-amino-2-phenylacetamide hydrochloride,l-phenylglycine amide hydrochloride,h-phg-nh2 hcl,h-phg-nh2.hcl,2s-2-amino-2-phenylacetamide hydrochloride,hpphg-nh2 hcl,h-phg-nh2 . hcl,phenylglycine amide hydrochloride,s-2-amino-2-phenyl-acetamide hydrochloride,alphas-alpha-amino-benzeneacetamide hydrochloride
22850726
(2S)-2-amino-2-phenylacetamide;hydrochloride
C1=CC=C(C=C1)C(C(=O)N)N.Cl
Quantity 5g
Beilstein 4823667
Formula Weight 186.64
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Phenyl-L-glycinamide hydrochloride