Alfa Aesar™ 2-Phenylanthraquinone, 98%

Manufacturer: Alfa Aesar™ H27376MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27376MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6485-97-8
    Molecular Formula C20H12O2
    Formula Weight 284.32
    MDL Number MFCD00559262
    Chemical Name or Material 2-Phenylanthraquinone

    Specifications

    Formula Weight 284.32
    Molecular Formula C20H12O2
    MDL Number MFCD00559262
    CAS 6485-97-8
    Melting Point 160° to 163°C
    Quantity 250mg
    Chemical Name or Material 2-Phenylanthraquinone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit