Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylbenzothiazole, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
883-93-2
C13H9NS
211.282
MFCD00005777
XBHOUXSGHYZCNH-UHFFFAOYSA-N
2-phenylbenzothiazole,2-phenylbenzo d thiazole,benzothiazole, 2-phenyl,unii-fo573g4bgt,2-phenyl benzothiazole,2-phenylbenzthiazole,fo573g4bgt,wln: t56 bn dsj cr,phenyl-benzothiazole,benzothiazole,phenyl
13439
2-phenyl-1,3-benzothiazole
C1=CC=C(C=C1)C2=NC3=CC=CC=C3S2
883-93-2
C13H9NS
211.282
MFCD00005777
XBHOUXSGHYZCNH-UHFFFAOYSA-N
2-phenylbenzothiazole,2-phenylbenzo d thiazole,benzothiazole, 2-phenyl,unii-fo573g4bgt,2-phenyl benzothiazole,2-phenylbenzthiazole,fo573g4bgt,wln: t56 bn dsj cr,phenyl-benzothiazole,benzothiazole,phenyl
13439
2-phenyl-1,3-benzothiazole
C1=CC=C(C=C1)C2=NC3=CC=CC=C3S2
Color Gray
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 111°C to 115°C
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 27, 74
Formula Weight 211.28
Physical Form Needle-Like Crystals
Percent Purity ≥96.0%
Chemical Name or Material 2-Phenylbenzothiazole, 97%
Show More Show Less