Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylbutyramide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
90-26-6
C10H13NO
163.22
MFCD00025511
UNFGQCCHVMMMRF-UHFFFAOYSA-N
2-phenylbutyramide,geristerol,hyposterol,normosterolo,phenetamid,phenetamide,redusterol,substerina,eusterol,geriapan
7011
2-phenylbutanamide
CCC(C1=CC=CC=C1)C(=O)N
90-26-6
C10H13NO
163.22
MFCD00025511
UNFGQCCHVMMMRF-UHFFFAOYSA-N
2-phenylbutyramide,geristerol,hyposterol,normosterolo,phenetamid,phenetamide,redusterol,substerina,eusterol,geriapan
7011
2-phenylbutanamide
CCC(C1=CC=CC=C1)C(=O)N
Quantity 25g
Color White
Melting Point 83°C to 87°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NCOCH(C2H5)C6H5
Merck Index 13, 7362
Formula Weight 163.22
Physical Form Crystalline Powder or Needles
Percent Purity ≥97.5%
Chemical Name or Material 2-Phenylbutyramide
Show More Show Less

EINECSNumber : 201-980-4

RTECSNumber : ES4955000

TSCA : TSCA