Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Phenylbutyramide, 98%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
Specifications
Packaging | Glass Bottle |
Quantity | 5g |
Infrared Spectrum | Authentic |
Color | White |
Melting Point | 83°C to 87°C |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 98% |
Linear Formula | H2NCOCH(C2H5)C6H5 |
Merck Index | 13, 7362 |
Show More |
Safety and Handling
EINECSNumber : 201-980-4
RTECSNumber : ES4955000
TSCA : TSCA