Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ (+/-)-2-Phenylbutyric acid, 98%
Click to view available options
Quantity:
100g
500g
Specifications
Refractive Index | 1.515 |
Quantity | 500g |
Boiling Point | 270°C to 272°C |
Melting Point | 40°C to 44°C |
Flash Point | >110°C (230°F) |
Beilstein | 509876 |
Formula Weight | 164.2 |
Percent Purity | 98% |
Chemical Name or Material | (+/-)-2-Phenylbutyric acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 201-982-5
RTECSNumber : ET5957500
TSCA : Yes
Recommended Storage : Ambient temperatures