Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Phenylbutyronitrile, 95%

Click to view available options
Quantity:
10g
50g
769-68-6
C10H11N
145.205
MFCD00001876
IZPUPXNVRNBDSW-UHFFFAOYSA-N
2-phenylbutyronitrile,2-phenylbutanentrile,1-cyano-1-phenylpropane,butanenitrile, 2-phenyl,benzeneacetonitrile, .alpha.-ethyl,butyronitrile, 2-phenyl,benzeneacetonitrile, ?-ethyl,.alpha.-phenylbutyronitrile,alpha-phenylbutyronitrile,dl-2-phenylbutyronitrile
95334
2-phenylbutanenitrile
CCC(C#N)C1=CC=CC=C1
769-68-6
C10H11N
145.205
MFCD00001876
IZPUPXNVRNBDSW-UHFFFAOYSA-N
2-phenylbutyronitrile,2-phenylbutanentrile,1-cyano-1-phenylpropane,butanenitrile, 2-phenyl,benzeneacetonitrile, .alpha.-ethyl,butyronitrile, 2-phenyl,benzeneacetonitrile, ?-ethyl,.alpha.-phenylbutyronitrile,alpha-phenylbutyronitrile,dl-2-phenylbutyronitrile
95334
2-phenylbutanenitrile
CCC(C#N)C1=CC=CC=C1
Density 0.975
Refractive Index 1.5086
Quantity 50g
Boiling Point 102°C to 104°C (7mmHg)
Flash Point 105°C (221°F)
UN Number UN3276
Formula Weight 145.2
Percent Purity 95%
Chemical Name or Material 2-Phenylbutyronitrile

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 212-213-8

TSCA : Yes

Recommended Storage : Ambient temperatures