Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylbutyryl chloride, 96%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
36854-57-6
C10H11ClO
182.647
MFCD00018811
QGXMHCMPIAYMGT-UHFFFAOYSA-N
2-phenylbutyryl chloride,2-ethyl-2-phenylacetyl chloride,2-phenylbutyrylchloride,butyryl chloride, 2-phenyl,acmc-209ipa,2-phenyl butyryl chloride,2-phenyl-butyryl chloride,2-phenyl-mutanoyl chloride,.alpha.-phenylbutyryl chloride,2-phenyl-2-ethylacetyl chloride
98173
2-phenylbutanoyl chloride
CCC(C1=CC=CC=C1)C(=O)Cl
36854-57-6
C10H11ClO
182.647
MFCD00018811
QGXMHCMPIAYMGT-UHFFFAOYSA-N
2-phenylbutyryl chloride,2-ethyl-2-phenylacetyl chloride,2-phenylbutyrylchloride,butyryl chloride, 2-phenyl,acmc-209ipa,2-phenyl butyryl chloride,2-phenyl-butyryl chloride,2-phenyl-mutanoyl chloride,.alpha.-phenylbutyryl chloride,2-phenyl-2-ethylacetyl chloride
98173
2-phenylbutanoyl chloride
CCC(C1=CC=CC=C1)C(=O)Cl
Density 1.093g/mL
Quantity 5g
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5160 to 1.5180 (20°C, 589nm)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula C6H5CH(CHCH3)C(O)Cl
Beilstein 09, IV, 1817
Formula Weight 182.65
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 2-Phenylbutyryl chloride
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 253-241-0

TSCA : TSCA