Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylcyclohexanone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1444-65-1
C12H14O
174.243
MFCD00001629
DRLVMOAWNVOSPE-UHFFFAOYSA-N
2-phenylcyclohexanone,cyclohexanone, 2-phenyl,2-phenyl-cyclohexanone,2-phenyl cyclohexanone,cyclohexanone, phenyl,2-phenylcyclohexanon,phenylcyclohexanone,phenyl cyclohexanone,6-phenylcyclohexanone
95592
2-phenylcyclohexan-1-one
C1CCC(=O)C(C1)C2=CC=CC=C2
1444-65-1
C12H14O
174.243
MFCD00001629
DRLVMOAWNVOSPE-UHFFFAOYSA-N
2-phenylcyclohexanone,cyclohexanone, 2-phenyl,2-phenyl-cyclohexanone,2-phenyl cyclohexanone,cyclohexanone, phenyl,2-phenylcyclohexanon,phenylcyclohexanone,phenyl cyclohexanone,6-phenylcyclohexanone
95592
2-phenylcyclohexan-1-one
C1CCC(=O)C(C1)C2=CC=CC=C2
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 56°C to 59°C
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5C6H9(=O)
Beilstein 07, I, 199
Formula Weight 174.24
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Phenylcyclohexanone, 98%
Show More Show Less