Learn More
2-Phenylethyl acetate, 98%, Thermo Scientific Chemicals
Description
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.
Solubility
It is insoluble in paraffin oil, water. Soluble in alcohol, dipropylene glycol, fixed oils, kerosene, propylene glycol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Protected from heat and light.
Specifications
Melting Point | -31°C |
Density | 1.029 |
Boiling Point | 231°C to 233°C |
Flash Point | 107°C (225°F) |
Odor | Fruit-like |
Refractive Index | 1.499 |
Quantity | 250 g |
Beilstein | 638179 |
Solubility Information | It is insoluble in paraffin oil,water. Soluble in alcohol,dipropylene glycol,fixed oils,kerosene,propylene glycol. |
Formula Weight | 164.21 |
Show More |
Safety and Handling
EINECSNumber | 203-113-5 |
RTECSNumber | AJ2220000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.