Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylglycinonitrile Hydrochloride (Tech.), 95%, Acros Organics

Click to view available options
Quantity:
25g
53941-45-0
C8H9ClN2
168.624
OGSYCVCLXAFDNE-DDWIOCJRSA-N
s-2-amino-2-phenylacetonitrile hydrochloride,alpha-phenylglycinonitrile hydrochloride,2-amino-2-phenyl-acetonitrile hydrochloride,2s-2-amino-2-phenylacetonitrile hydrochloride
22870696
(2S)-2-amino-2-phenylacetonitrile;hydrochloride
C1=CC=C(C=C1)C(C#N)N.Cl
53941-45-0
C8H9ClN2
168.624
OGSYCVCLXAFDNE-DDWIOCJRSA-N
s-2-amino-2-phenylacetonitrile hydrochloride,alpha-phenylglycinonitrile hydrochloride,2-amino-2-phenyl-acetonitrile hydrochloride,2s-2-amino-2-phenylacetonitrile hydrochloride
22870696
(2S)-2-amino-2-phenylacetonitrile;hydrochloride
C1=CC=C(C=C1)C(C#N)N.Cl
Quantity 25g
Color Yellow to Brown
Infrared Spectrum Authentic
Melting Point 158°C to 165°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C6H5CH(NH2)CN·HCl
Beilstein 14, 462
Formula Weight 168.63
Physical Form Powder or Chunks
Percent Purity ≥94%
Grade Technical
Chemical Name or Material 2-Phenylglycinonitrile hydrochloride, tech., 95%
Show More Show Less