Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Phenylimidazole, 98+%

Manufacturer:  Alfa Aesar™ B2385822

Quantity 100g

 View more versions of this product

Catalog No. AAB2385822


  Check Availability
Add to cart

Specifications

Description

Applications
2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.

Solubility
Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

670-96-2
144.18
114011
C9H8N2
MFCD00005186
2-Phenylimidazole

Specifications

200°C (392°F)
146° to 150°C
670-96-2
144.18
114011
≥98%
198° to 200°C (7mmHg)
100g
C9H8N2
MFCD00005186
2-Phenylimidazole

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit