Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylimidazole, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
670-96-2
C9H8N2
144.18
MFCD00005186
ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
2-phenylimidazole,1h-imidazole, 2-phenyl,imidazole, 2-phenyl,2-phenyl imidazole,2-phenyl-1h-imidazol,unii-7r2590hm4l,chembl14189,phenyl imidazole,2-phenyl-imidazole,piy
69591
2-phenyl-1H-imidazole
C1=CC=C(C=C1)C2=NC=CN2
670-96-2
C9H8N2
144.18
MFCD00005186
ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
2-phenylimidazole,1h-imidazole, 2-phenyl,imidazole, 2-phenyl,2-phenyl imidazole,2-phenyl-1h-imidazol,unii-7r2590hm4l,chembl14189,phenyl imidazole,2-phenyl-imidazole,piy
69591
2-phenyl-1H-imidazole
C1=CC=C(C=C1)C2=NC=CN2
Melting Point 141.0°C to 148.0°C
Quantity 25g
Boiling Point 340.0°C
Color Yellow to Beige
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 200°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 23, 182
Solubility Information Solubility in water: insoluble
Formula Weight 144.18
Physical Form Powder, Granular or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Phenylimidazole, 95%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-581-7

TSCA : TSCA