Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylimidazole, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
670-96-2
C9H8N2
144.18
MFCD00005186
ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
2-phenylimidazole,1h-imidazole, 2-phenyl,imidazole, 2-phenyl,2-phenyl imidazole,2-phenyl-1h-imidazol,unii-7r2590hm4l,chembl14189,phenyl imidazole,2-phenyl-imidazole,piy
69591
2-phenyl-1H-imidazole
C1=CC=C(C=C1)C2=NC=CN2
670-96-2
C9H8N2
144.18
MFCD00005186
ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
2-phenylimidazole,1h-imidazole, 2-phenyl,imidazole, 2-phenyl,2-phenyl imidazole,2-phenyl-1h-imidazol,unii-7r2590hm4l,chembl14189,phenyl imidazole,2-phenyl-imidazole,piy
69591
2-phenyl-1H-imidazole
C1=CC=C(C=C1)C2=NC=CN2
Melting Point 141.0°C to 148.0°C
Boiling Point 340.0°C
Flash Point 200°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow to Beige
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 23, 182
Solubility Information Solubility in water: insoluble
Formula Weight 144.18
Physical Form Powder, Granular or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Phenylimidazole, 95%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 211-581-7

TSCA : TSCA