Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylindole 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
Plastic bottle
948-65-2
C14H11N
193.25
MFCD00005608
KLLLJCACIRKBDT-UHFFFAOYSA-N
2-phenylindole,1h-indole, 2-phenyl,indole, 2-phenyl,stabilizer i,alpha-phenylindole,2-phenyl indole,phenylindole,unii-mqd44hv3p1,.alpha.-phenylindole,mqd44hv3p1
13698
2-phenyl-1H-indole
C1=CC=C(C=C1)C2=CC3=CC=CC=C3N2
948-65-2
C14H11N
193.25
MFCD00005608
KLLLJCACIRKBDT-UHFFFAOYSA-N
2-phenylindole,1h-indole, 2-phenyl,indole, 2-phenyl,stabilizer i,alpha-phenylindole,2-phenyl indole,phenylindole,unii-mqd44hv3p1,.alpha.-phenylindole,mqd44hv3p1
13698
2-phenyl-1H-indole
C1=CC=C(C=C1)C2=CC3=CC=CC=C3N2
Color Beige to White or Green
Boiling Point 250.0°C (10.0 mmHg)
Sulfated Ash 0.2% max.
Packaging Plastic bottle
Melting Point 188.0°C to 191.0°C
Infrared Spectrum Authentic
Quantity 500g
Water 0.5% max. (K.F.)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, 467
Solubility Information Solubility in water: .. Other solubilities: soluble in thf and acetone,slightly soluble in methanol
Formula Weight 193.25
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 2-Phenylindole, 99%
Show More Show Less

EINECSNumber : 213-436-3

RTECSNumber : NM1272500

TSCA : TSCA