Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylisobutyric Acid 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
826-55-1
C10H12O2
164.20
MFCD00014332
YYEROYLAYAVZNW-UHFFFAOYSA-N
2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
13222
2-methyl-2-phenylpropanoic acid
CC(C)(C(O)=O)C1=CC=CC=C1
826-55-1
C10H12O2
164.20
MFCD00014332
YYEROYLAYAVZNW-UHFFFAOYSA-N
2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
13222
2-methyl-2-phenylpropanoic acid
CC(C)(C(O)=O)C1=CC=CC=C1
Color White-Yellow
Melting Point 79°C
Quantity 250g
Formula Weight 164.20
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Phenylisobutyric Acid